Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Digestion Standard Mix, Mix 1, Tryptically Digested, Equimolar Ratio

Catalog No. 50781408 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

MassPREP™ Protein Digestion Standard Mix 1 consists of a relatively close equimolar ratio of tryptically digested proteins. This standard is used for verification of Waters PLGS software.

  • Consists of a relatively close equimolar ratio of tryptically digested proteins: yeast enolase (SwissProt P00924), phosphorylase b (SwissProt P00489), yeast alcohol dehydrogenase (ADH, SwissProt P00330) and bovine serum albumin (BSA, SwissProt P02769).
  • Exact ratio is published on the certificate of analysis (COA).
  • Used for verification of PLGS software.
  • Separation mode: reversed phase.
  • Detection method: UV.
  • Formulation: solid.
  • Storage condition: room temperature.
  • System type: LC.
  • Shelf life: 60 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: ambient.

Catalog No. 50-781-408 Supplier Waters Corp Supplier No. 186002865
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MassPREP™ Protein Digestion Standard Mix 1 consists of a relatively close equimolar ratio of tryptically digested proteins. This standard is used for verification of Waters PLGS software.

  • Consists of a relatively close equimolar ratio of tryptically digested proteins: yeast enolase (SwissProt P00924), phosphorylase b (SwissProt P00489), yeast alcohol dehydrogenase (ADH, SwissProt P00330) and bovine serum albumin (BSA, SwissProt P02769).
  • Exact ratio is published on the certificate of analysis (COA).
  • Used for verification of PLGS software.
  • Separation mode: reversed phase.
  • Detection method: UV.
  • Formulation: solid.
  • Storage condition: room temperature.
  • System type: LC.
  • Shelf life: 60 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: ambient.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.