Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Protein Standard Mix, Reversed phase, UV detection, Solid

Catalog No. NC9031250 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The MassPREP Protein Standard Mix is composed of Ribonuclease A, Cytochrome c, bovine serum Albumin, Myoglobin, Enolase, and Phosphorylase b. This mixture offers a variety of proteins with different isoelectric points, molecular weights, and hydrophobicities, making it suitable for various applications. It is recommended for regular use to benchmark column and system performance, similar to the QCRM product line.

  • Separation mode: Reversed phase
  • Detection method: UV
  • Formulation: Solid
  • Storage condition: -20°C (Freezer)
  • System type: LC
  • Shelf life: 24 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Mass: ~ 123 μg/vial
  • Type: Reference
  • Application: Protein

Catalog No. NC9031250 Supplier Waters Corp Supplier No. 186004900
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MassPREP Protein Standard Mix is composed of Ribonuclease A, Cytochrome c, bovine serum Albumin, Myoglobin, Enolase, and Phosphorylase b. This mixture offers a variety of proteins with different isoelectric points, molecular weights, and hydrophobicities, making it suitable for various applications. It is recommended for regular use to benchmark column and system performance, similar to the QCRM product line.

  • Separation mode: Reversed phase
  • Detection method: UV
  • Formulation: Solid
  • Storage condition: -20°C (Freezer)
  • System type: LC
  • Shelf life: 24 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Mass: ~ 123 μg/vial
  • Type: Reference
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.