Learn More
Waters Corp MassTrak Endocrine Steroid Calibrator Set
Catalog No. 502269317 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassTrak Endocrine Steroid Calibrator Set is intended to be used in a clinical biochemistry laboratory by trained professionals for the quantitative determination of steroids (mineralocorticoids, androgens, glucocorticoids and progestogens) in human serum to aid diagnosis of steroid related disorders. The MassTrak Endocrine Steroid Calibrator Set are used to establish a calibration curve which is used to interpolate the steroid concentrations using liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.The Waters MassTrak Endocrine Steroid Calibrator Set [one box] contains 7 matrix-matched calibrators in human serum and labeling. Contents include:• Calibrator 0 (Cal. 0)• Calibrator 1 (Cal. 1)• Calibrator 2 (Cal. 2)• Calibrator 3 (Cal. 3)• Calibrator 4 (Cal. 4)• Calibrator 5 (Cal. 5)• Calibrator 6 (Cal. 6)
This item is not returnable. View return policy
Catalog No. 50-226-9317 Supplier Waters Corp Supplier No. 186009311IVD
The MassTrak Endocrine Steroid Calibrator Set is intended to be used in a clinical biochemistry laboratory by trained professionals for the quantitative determination of steroids (mineralocorticoids, androgens, glucocorticoids and progestogens) in human serum to aid diagnosis of steroid related disorders. The MassTrak Endocrine Steroid Calibrator Set are used to establish a calibration curve which is used to interpolate the steroid concentrations using liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.The Waters MassTrak Endocrine Steroid Calibrator Set [one box] contains 7 matrix-matched calibrators in human serum and labeling. Contents include:• Calibrator 0 (Cal. 0)• Calibrator 1 (Cal. 1)• Calibrator 2 (Cal. 2)• Calibrator 3 (Cal. 3)• Calibrator 4 (Cal. 4)• Calibrator 5 (Cal. 5)• Calibrator 6 (Cal. 6)