Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MassTrak Immunosuppressant Calibrator Set, LCMS, 3 mL

Catalog No. 502269340 Shop All Waters Corporation Products
Encompass_Preferred

The MassTrak Immunosuppressant Calibrator Set offers seven matrix-matched, lyophilized calibrators in pooled human whole blood. Designed for clinical laboratories, it enables accurate quantification of cyclosporine, sirolimus, tacrolimus, and everolimus in human whole blood samples. This set aids in managing immunosuppressive drug therapy by establishing a calibration curve via liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.

  • Seven matrix-matched calibrators.
  • Contains everolimus, cyclosporine, tacrolimus, and sirolimus.
  • Detection method: MS.
  • System type: LCMS.
  • Quantitative product.
  • Shelf life: 60 months (unopened).
  • Shelf life upon reconstitution: 1 month.
  • Requires reconstitution with 3 mL of LC-MS-grade water.
  • Traceability assessed by ISO17511 (2020).

This item is not returnable. View return policy
Catalog No. 50-226-9340 Supplier Waters Corp Supplier No. 186010251IVD
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MassTrak Immunosuppressant Calibrator Set offers seven matrix-matched, lyophilized calibrators in pooled human whole blood. Designed for clinical laboratories, it enables accurate quantification of cyclosporine, sirolimus, tacrolimus, and everolimus in human whole blood samples. This set aids in managing immunosuppressive drug therapy by establishing a calibration curve via liquid chromatography – tandem mass spectrometry (LC-MS/MS) systems.

  • Seven matrix-matched calibrators.
  • Contains everolimus, cyclosporine, tacrolimus, and sirolimus.
  • Detection method: MS.
  • System type: LCMS.
  • Quantitative product.
  • Shelf life: 60 months (unopened).
  • Shelf life upon reconstitution: 1 month.
  • Requires reconstitution with 3 mL of LC-MS-grade water.
  • Traceability assessed by ISO17511 (2020).

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.