Learn More
Waters Corp Steroid Serum Calibrator Set, Set 2, RUO, NIST Traceable
Catalog No. 502392977 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This calibrator set is designed for quantifying steroids in human serum for clinical research methods. The calibrators are traceable to NIST CRM and reference measurement procedures like RfB (Germany) and the University of Ghent for selected analytes, offering customer-guided wide concentration ranges for all analytes.
  • Ideal for quantifying steroids in human serum for clinical research methods.
  • Calibrators are traceable to NIST CRM and reference measurement procedures such as RfB (Germany) and University of Ghent for select analytes.
  • Customer-guided wide concentration ranges for all analytes.
Catalog No. 50-239-2977 Supplier Waters Corp Supplier No. 186010563
This calibrator set is designed for quantifying steroids in human serum for clinical research methods. The calibrators are traceable to NIST CRM and reference measurement procedures like RfB (Germany) and the University of Ghent for selected analytes, offering customer-guided wide concentration ranges for all analytes.
  • Ideal for quantifying steroids in human serum for clinical research methods.
  • Calibrators are traceable to NIST CRM and reference measurement procedures such as RfB (Germany) and University of Ghent for select analytes.
  • Customer-guided wide concentration ranges for all analytes.