Learn More
Waters Corp MassTrak Steroid Serum QC Set 3, RUO, LCMS, 2 mL, 3 Year Shelf Life
Catalog No. 502392061 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Steroid Serum QC Set 3 is for Research Use Only (RUO), providing serum-based lyophilized quality controls for 5 steroids at 4 concentrations. It offers traceability to NIST CRM and other reference measurement procedures, designed for optimal clinical research method performance in quantifying steroid hormones in serum by LC-MS/MS.
  • Designed for LCMS detection.
  • Solid formulation.
  • Shelf life of 3 years.
  • Offers in-use stability for 1 month at 2–8°C and 3 months at ≤-10°C.
  • Reconstituted vial volume is 2 mL.
  • Saves up to 3 months of skilled user time during method development.
  • Supports laboratory compliance to ISO 15189 and enables longitudinal result harmonization.
Catalog No. 50-239-2061 Supplier Waters Corp Supplier No. 186010566
This Steroid Serum QC Set 3 is for Research Use Only (RUO), providing serum-based lyophilized quality controls for 5 steroids at 4 concentrations. It offers traceability to NIST CRM and other reference measurement procedures, designed for optimal clinical research method performance in quantifying steroid hormones in serum by LC-MS/MS.
  • Designed for LCMS detection.
  • Solid formulation.
  • Shelf life of 3 years.
  • Offers in-use stability for 1 month at 2–8°C and 3 months at ≤-10°C.
  • Reconstituted vial volume is 2 mL.
  • Saves up to 3 months of skilled user time during method development.
  • Supports laboratory compliance to ISO 15189 and enables longitudinal result harmonization.