Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp QC Standards, Steroid Serum, 2 mL, LCMS, RUO

Catalog No. 502390678 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This QC set contains serum–based lyophilized quality controls that are traceable to NIST CRM and reference measurement procedures such as RfB (Germany) and University of Ghent for select analytes. The concentration values for the controls are informed by customer feedback, Endocrine Society guidelines, and CDC NSQAP. It is intended for Research Use Only.

  • Detection method: LCMS.
  • Solid formulation.
  • Storage condition: –20°C ± 10°C (Freezer).
  • Quantitative product.
  • Shelf life: 3 years at ≤–10°C.
  • In-use stability: 1 month at 2–8°C, 3 months at ≤–10°C.
  • Traceable to NIST.
  • Shipping condition: Ambient.
  • Volume: 2 mL.
  • One unit per package.
  • Contains serum–based lyophilized quality controls for 4 steroids at 4 different concentration values.
  • Includes an insert card and the CoA.

Catalog No. 50-239-0678 Supplier Waters Corp Supplier No. 186010564
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This QC set contains serum–based lyophilized quality controls that are traceable to NIST CRM and reference measurement procedures such as RfB (Germany) and University of Ghent for select analytes. The concentration values for the controls are informed by customer feedback, Endocrine Society guidelines, and CDC NSQAP. It is intended for Research Use Only.

  • Detection method: LCMS.
  • Solid formulation.
  • Storage condition: –20°C ± 10°C (Freezer).
  • Quantitative product.
  • Shelf life: 3 years at ≤–10°C.
  • In-use stability: 1 month at 2–8°C, 3 months at ≤–10°C.
  • Traceable to NIST.
  • Shipping condition: Ambient.
  • Volume: 2 mL.
  • One unit per package.
  • Contains serum–based lyophilized quality controls for 4 steroids at 4 different concentration values.
  • Includes an insert card and the CoA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.