Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp MassTrak Endocrine Blank Calibrator, Charcoal Stripped Serum, LCMS, Clinical Diagnostics

Catalog No. 502886222 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Endocrine Blank Calibrator is prepared with charcoal stripped serum, stripped 6 times, and is intended for use in Clinical Diagnostics and Endocrinology applications.

  • Prepared with charcoal stripped serum, stripped 6 times.
  • Intended for clinical diagnostics and endocrinology applications.
  • Detection method is LCMS.
  • Formulation is solid.
  • Storage condition is −20°C ± 10°C (Freezer).
  • Quantitative.
  • Shelf life of 36 months.
  • Shelf life upon reconstitution is 1 month.
  • NIST traceable.
  • Ships ambient.
  • Volume of 2 mL.
  • Product type is standards.
  • One unit per package.

Catalog No. 50-288-6222 Supplier Waters Corp Supplier No. 186010560IVD
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Endocrine Blank Calibrator is prepared with charcoal stripped serum, stripped 6 times, and is intended for use in Clinical Diagnostics and Endocrinology applications.

  • Prepared with charcoal stripped serum, stripped 6 times.
  • Intended for clinical diagnostics and endocrinology applications.
  • Detection method is LCMS.
  • Formulation is solid.
  • Storage condition is −20°C ± 10°C (Freezer).
  • Quantitative.
  • Shelf life of 36 months.
  • Shelf life upon reconstitution is 1 month.
  • NIST traceable.
  • Ships ambient.
  • Volume of 2 mL.
  • Product type is standards.
  • One unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.