Learn More
Bachem Mast Cell Degranulating (MCD) Peptide
SDP
Supplier:  Bachem 4030775.0500
Encompass_Preferred
Mast Cell Degranulating (MCD) Peptide, H-Ile-Lys-Cys-Asn-Cys-Lys-Arg-His-Val-Ile-Lys-Pro-His-Ile-Cys-Arg-Lys-Ile-Cys-Gly-Lys-Asn-NH2, 32908-73-9, C110H192N40O24S4, Mr 2587.25, 0.5mg. Peptide 401. The highly basic MCD peptide is present in small amounts in the venom of the European honeybee, Apis mellifera. At low doses, it causes mast cell degranulation and subsequent histamine release. At higher doses, it produces an anti-inflammatory effect, which may be beneficial in rheumatic and allergic diseases.
Catalog No. 50-383-387