Learn More
CPC Scientific H-Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MCDP001A
Encompass
MCD Peptide HR-1 is isolated from the venom of the giant hornet Vespa Orientalis and differs structurally from the MCD peptide from honeybee venom but possesses similar biological effects. These peptides are capable of degranulating rat mast cells, thereby initiating histamine release. ONE-LETTER SEQUENCE: INLKAIAALVKKVL-NH2MOLECULAR FORMULA: C71H133N19O15MOLECULAR WEIGHT:1492.96STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [80533-94-4], SYNONYMS: Mast Cell Degranulating (MCD) Peptide HR-1RESEARCH AREA: InflammationREFERENCES: M.U. Tuichibaev et al., Biochem. USSR, 53, 183 (1988)
Catalog No. 50-210-9713