Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mast cell degranulating peptide | 32908-73-9 | 98.3% | 2,587.22 g/mol | C110H192N40O24S4 | 5MG
SDP

Catalog No. 5000235694
Encompass_Preferred

Peptide 401 is a mast cell-degranulating peptide derived from bee venom, supplied lyophilized for research use. It is suitable for studies of mast cell activation, histamine release, and vascular permeability in cellular and animal models, and is provided in small pack sizes for experimental work.

  • Sequence: Ile-Lys-Cys-Asn-Cys-Lys-Arg-His-Val-Ile-Lys-Pro-His-Ile-Cys-Arg-Lys-Ile-Cys-Gly-Lys-Asn-NH2.
  • Molecular weight: 2587.22 g/mol.
  • Chemical formula: C110H192N40O24S4.
  • Purity: 98.3%.
  • Solubility: water 2.08 mg/mL; ultrasonic assistance recommended.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 1 mg, 5 mg, 10 mg.

Catalog No. 50-002-35694 Supplier Medchemexpress LLC Supplier No. HY125375MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peptide 401 is a mast cell-degranulating peptide derived from bee venom, supplied lyophilized for research use. It is suitable for studies of mast cell activation, histamine release, and vascular permeability in cellular and animal models, and is provided in small pack sizes for experimental work.

  • Sequence: Ile-Lys-Cys-Asn-Cys-Lys-Arg-His-Val-Ile-Lys-Pro-His-Ile-Cys-Arg-Lys-Ile-Cys-Gly-Lys-Asn-NH2.
  • Molecular weight: 2587.22 g/mol.
  • Chemical formula: C110H192N40O24S4.
  • Purity: 98.3%.
  • Solubility: water 2.08 mg/mL; ultrasonic assistance recommended.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solution -80°C (6 months) or -20°C (1 month).
  • Available pack sizes: 1 mg, 5 mg, 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.