Learn More
Biolegend Purified anti-human Mast Cell Tryptase
Supplier:  Biolegend 369402
Encompass_Preferred
Purified anti-human Mast Cell Tryptase [AA1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Monkey, Sheep (Ovine), Pig (Swine, Porcine), Cow (Cattle, Bovine), Dog (Canine), Cat (Feline), Horse (Equine); Apps: IHC-P, ICFC, ELISA, WB; Size: 100 ug
Catalog No. NC1661714