Learn More
Enterprise Technology Solutions Master Caster Casters w/ Brake, J-Plate Stem, 3" Diameter, 4/ST, GY/BK
SDP
Catalog No. 5000791564
Encompass_Preferred
3" Gemini casters with a J-plate feature a decorative, two-toned body to provide a modern look for a variety of your equipment. Toggle brakes quickly lock and release to prevent rolling on smooth surfaces. High weight rating safely supports up to 220 lb. per wheel. Soft, twin-wheel casters are safe for use on hard floors and chairmats. Gemini casters are perfect for adding mobility to cabinets, files, tables, chairs and more.
Catalog No. 50-007-91564 Supplier Enterprise Technology Solutions Supplier No. MAS71444
3" Gemini casters with a J-plate feature a decorative, two-toned body to provide a modern look for a variety of your equipment. Toggle brakes quickly lock and release to prevent rolling on smooth surfaces. High weight rating safely supports up to 220 lb. per wheel. Soft, twin-wheel casters are safe for use on hard floors and chairmats. Gemini casters are perfect for adding mobility to cabinets, files, tables, chairs and more.