Learn More
Enterprise Technology Solutions Master Caster Furniture Repair Kit,5 Touch up Markers,3 Filler Sticks,AST
SDP
Catalog No. 5000791848
Encompass_Preferred
Furniture Repair Kit repairs and restores color to any wood surface. Use on furniture, floors, woodwork, cabinets, doors, paneling and frames. Conceals minor scratches and worn edges. Repairs easily with no experience required. Markers seal, stain and finish in one easy application. Kit includes five woodgrain markers (for use with natural, light oak, dark oak, cherry and walnut and mahogany finishes) and three filler sticks (light, medium and dark).
Catalog No. 50-007-91848 Supplier Enterprise Technology Solutions Supplier No. MAS18000
Furniture Repair Kit repairs and restores color to any wood surface. Use on furniture, floors, woodwork, cabinets, doors, paneling and frames. Conceals minor scratches and worn edges. Repairs easily with no experience required. Markers seal, stain and finish in one easy application. Kit includes five woodgrain markers (for use with natural, light oak, dark oak, cherry and walnut and mahogany finishes) and three filler sticks (light, medium and dark).