Learn More
Enterprise Technology Solutions Master Caster Futura Caster, B Stem, Soft Wheel, 7/16"Dx7/8"H, 5/ST,BK
SDP
Catalog No. 5000791707
Encompass_Preferred
Futura dual wheel caster is designed exclusively for "Euro-style" chairs. Specially designed 1-1/2" diameter neck fits 1-1/2" diameter chair bases, creating the illusion of chair leg and caster being one unit. 2-3/16" diameter, dual-wheel soft caster is designed for use on hard floors and chairmats. Weight rating is 100 lb. per caster.
Catalog No. 50-007-91707 Supplier Enterprise Technology Solutions Supplier No. MAS94326
Futura dual wheel caster is designed exclusively for "Euro-style" chairs. Specially designed 1-1/2" diameter neck fits 1-1/2" diameter chair bases, creating the illusion of chair leg and caster being one unit. 2-3/16" diameter, dual-wheel soft caster is designed for use on hard floors and chairmats. Weight rating is 100 lb. per caster.