Learn More
Enterprise Technology Solutions Master Caster Giant Foot Doorstop, 3-1/2"x6-3/4"x2", Safety Yellow
SDP
Catalog No. 5000790048
Encompass_Preferred
Giant doorstop is designed to hold heavy, commercial and industrial doors and doors with automatic closures or spring closures and/or extra-high clearances up to 2". Large footprint grips tightly and stays in place when used on rough or uneven surfaces. Impact-resistant rubberized material stands up to daily use. Highly visible color helps prevent tripping in small spaces or doorways. Doorstop will not mar floors or carpets.
Catalog No. 50-007-90048 Supplier Enterprise Technology Solutions Supplier No. MAS00966
Giant doorstop is designed to hold heavy, commercial and industrial doors and doors with automatic closures or spring closures and/or extra-high clearances up to 2". Large footprint grips tightly and stays in place when used on rough or uneven surfaces. Impact-resistant rubberized material stands up to daily use. Highly visible color helps prevent tripping in small spaces or doorways. Doorstop will not mar floors or carpets.