Learn More
Electron Microscopy Sciences Master Coiler Pliers, 5.75"
SDP
Catalog No. 5033122
Encompass
This ʽMaster Coiler' is the painless pliers. It is designed to make loops and coils without nicking precious wire. Master Coiler allows one to hold the wire with the flat jaw while rotating the round jaw to form the perfect loop. Comfort grips, non-rust, highly polished stainless steel jaws and standard step-jaw for consistent loops and coils.
Catalog No. 50-331-22 Supplier Electron Microscopy Sciences Supplier No. 6218620
This ʽMaster Coiler' is the painless pliers. It is designed to make loops and coils without nicking precious wire. Master Coiler allows one to hold the wire with the flat jaw while rotating the round jaw to form the perfect loop. Comfort grips, non-rust, highly polished stainless steel jaws and standard step-jaw for consistent loops and coils.