Learn More
AVANTOR MASTERFLEX (VWR LLC) Cartridge pump head, L/S 8-channel, 3-roller.
Catalog No. NC1114557
Encompass
Masterflex L/S 8-channel, 3-roller cartridge pump head. Synchronous, multichannel flow from up to 8 channelsFlow rates: 0.074 to 1700 mL/minNumber of cartridges: 1 to 8 small, 1 to 4 large. Mix large and small cartridges¿one large cartridge occupies the same space as two small cartridgesGear ratio: 1:1Occlusion: tangential, adjustableDrive requirements: 1/10-hp min, 600 rpm max
Catalog No. NC1114557 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0751905
Masterflex L/S 8-channel, 3-roller cartridge pump head. Synchronous, multichannel flow from up to 8 channelsFlow rates: 0.074 to 1700 mL/minNumber of cartridges: 1 to 8 small, 1 to 4 large. Mix large and small cartridges¿one large cartridge occupies the same space as two small cartridgesGear ratio: 1:1Occlusion: tangential, adjustableDrive requirements: 1/10-hp min, 600 rpm max