Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Chem-Durance Bio, L/S 18; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644218/DEL
Encompass
Applications: Laboratory, pharmaceutical processing, and biotech Characteristics: Excellent chemical resistance, excellent life and durability under pressure, low spallation, plasticizer-free inner liner with elastomer jacket, opaque
Catalog No. NC9921396