Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled High-Performance Precision Pump Tubing, Peroxide-Cured Silicone, L/S 24; 200 ft
Catalog No. NC0063029
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC0063029 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640724
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.