Learn More
Enterprise Technology Solutions Master Lock Company Safety Padlock, Labeled, 1/4"Dx1-1/2"H Shackle, 6/PK, Red
SDP
Catalog No. 5000828476
Encompass_Preferred
Labeled safety padlock is ideal for lockouts and meets OSHA 1910.147 standard. Durable, nonconductive padlock is light and easy to carry. Key-retaining feature ensures that padlock is not left unlocked. Padlock reads "Danger. Locked Out. Do Not Remove." and has a six-pin, tumbler cylinder that is keyed differently.
Catalog No. 50-008-28476 Supplier Enterprise Technology Solutions Supplier No. MLK410REDPK
Labeled safety padlock is ideal for lockouts and meets OSHA 1910.147 standard. Durable, nonconductive padlock is light and easy to carry. Key-retaining feature ensures that padlock is not left unlocked. Padlock reads "Danger. Locked Out. Do Not Remove." and has a six-pin, tumbler cylinder that is keyed differently.