Learn More
Enterprise Technology Solutions Master Lock Company Warded Padlock, Durable Laminated Steel Body
SDP
Catalog No. 5000827846
Encompass_Preferred
Padlock features a durable laminated steel 1-1/2" (38mm) body. Warded locking mechanism minimizes jamming from dirt and grime. Mechanism tolerances make warded locks a frequent choice where sand, ice and other contaminants may jam other types of locks. Steel shackle provides strong cut-resistance. Includes pair of nickel-plated steel keys.
Catalog No. 50-008-27846 Supplier Enterprise Technology Solutions Supplier No. MLK22D
Padlock features a durable laminated steel 1-1/2" (38mm) body. Warded locking mechanism minimizes jamming from dirt and grime. Mechanism tolerances make warded locks a frequent choice where sand, ice and other contaminants may jam other types of locks. Steel shackle provides strong cut-resistance. Includes pair of nickel-plated steel keys.