Learn More
StatLab Master Tech Modified Mayer's Hematoxylin
SDP
Catalog No. 21033379
Change view
Click to view available options
Product Type:
Mayers Hematoxylin Stain
Catalog No. Product Type
21-033-379 Mayers Hematoxylin Stain
1 options
Catalog No. 21-033-379 Supplier StatLab Supplier No. HXMMHGAL
Add to Cart
Add to Cart
This modified version of Mayer’s formulation offers distinct and striking nuclear detail
  • A light delicate hematoxylin with no mucin affinity, it requires no differentiating and no bluing step to achieve optimal results
  • Ready-to-use, no filtering is required
Specifications
Product Type Mayers Hematoxylin Stain