Learn More
LGC GENOMICS LLC (Lucigen) MasterPure Yeast RNA Purification Kit, 100 PurificationsSafe, fast purification of high quality RNA from multiple species of yeast.
Catalog No. NC9520464
Encompass
The MasterPure™ Yeast RNA Purification Kit provides all of the reagents needed to purify RNA from yeast cell types (including Candida, Saccharomyces, Schizosaccharomyces) and filamentous fungi.
This item is not returnable. View return policy
Catalog No. NC9520464 Supplier LGC GENOMICS LLC Supplier No. MPY03100
The MasterPure™ Yeast RNA Purification Kit provides all of the reagents needed to purify RNA from yeast cell types (including Candida, Saccharomyces, Schizosaccharomyces) and filamentous fungi.