Learn More
LGC GENOMICS LLC (Lucigen) MasterAmp Tth DNA Polymerase, 250 U @ 5 U/LA thermostable DNA- and RNA-dependent DNA polymerase for both PCR and RT-PCR applications.
Catalog No. NC2162082
Encompass
Flexible: Use a single enzyme for both PCR and RT-PCR applications, greatly simplifying reaction optimization and setupMasterAmp Tth DNA Polymerase (5 U/L, 250 Units) is provided with MasterAmp Tth 20X PCR Buffer, MasterAmp 10X PCR Enhancer (with betaine), and 25 mM MgCl2, and MnSO4 Solutions to allow optimization of individual template-primer pair combination
This item is not returnable. View return policy
Catalog No. NC2162082 Supplier LGC GENOMICS LLC Supplier No. TTH72250
Flexible: Use a single enzyme for both PCR and RT-PCR applications, greatly simplifying reaction optimization and setupMasterAmp Tth DNA Polymerase (5 U/L, 250 Units) is provided with MasterAmp Tth 20X PCR Buffer, MasterAmp 10X PCR Enhancer (with betaine), and 25 mM MgCl2, and MnSO4 Solutions to allow optimization of individual template-primer pair combination