Learn More
USA Scientific Inc Mastercycler X50l
Catalog No. 502745495
Encompass
Mastercycler X50l, 110-230 V/50-60 Hz, aluminium block, 96-well or 0.1/0.2 mL tubes, eco unit. Eco units require being networked with a touchscreen model.
Catalog No. 50-274-5495 Supplier USA Scientific Inc Supplier No. 40630310
Add to Cart
Add to Cart
Request a Quote
Mastercycler X50l, 110-230 V/50-60 Hz, aluminium block, 96-well or 0.1/0.2 mL tubes, eco unit. Eco units require being networked with a touchscreen model.