Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex C/L Microflex Digital Pump with Single-Channel Pump Head for Microbore Tubing, 300 rpm
Catalog No. NC1500636
Encompass
This compact peristaltic pump features a durable, stackable ABS housing that is easy to wipe down and keep clean. The Microflex pump head features three rollers and a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.
Catalog No. NC1500636 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7712300
This compact peristaltic pump features a durable, stackable ABS housing that is easy to wipe down and keep clean. The Microflex pump head features three rollers and a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.