Learn More
AVANTOR MASTERFLEX (VWR LLC) Digital Process Drive w/ Stainless Steel Housing; 650 rpm
Catalog No. NC0618235
Encompass
Masterflex I/P Digital Process Drive with Stainless Steel Housing; 650 rpm. Precision digital drives offer washdown capability, programmable dispensing, and accurate low-maintenance motorsMinimum flow rate is 0.0006 LPM (0.0001 GPM); maximum flow rate is 26 LPM (6.9 GPM) with two stacked I/P Easy-Load or Standard pump headsHousings are NEMA 4X (IP66) rated for washdownConstructed with a durable 316 stainless steel housing with an anodized aluminum base
Catalog No. NC0618235 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7742010
Add to Cart
Add to Cart
Request a Quote
Masterflex I/P Digital Process Drive with Stainless Steel Housing; 650 rpm. Precision digital drives offer washdown capability, programmable dispensing, and accurate low-maintenance motorsMinimum flow rate is 0.0006 LPM (0.0001 GPM); maximum flow rate is 26 LPM (6.9 GPM) with two stacked I/P Easy-Load or Standard pump headsHousings are NEMA 4X (IP66) rated for washdownConstructed with a durable 316 stainless steel housing with an anodized aluminum base