Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Head for High-Performance Precision Tubing
Catalog No. NC1401416
Encompass
The L/S High-Performance pump heads offer better suction lift, viscous fluid transfer, and longer tubing life. Tubing enters and exits the same side of the pump head for easy connection to tubing inlet and outlet. Heads are not compatible with the 07554-series L/S Economy Console drives.
Catalog No. NC1401416 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7725272
The L/S High-Performance pump heads offer better suction lift, viscous fluid transfer, and longer tubing life. Tubing enters and exits the same side of the pump head for easy connection to tubing inlet and outlet. Heads are not compatible with the 07554-series L/S Economy Console drives.