Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Fixed-Speed Drive, 20 rpm; 115 VAC
Catalog No. NC9745852
Encompass
This is an economical speed drive with seven speeds for reliable general transfer and feed applications. The drive is stackable and ideal for lab, field or process feed applications. The drive is IP22-rated and protected from water falling vertically.
Catalog No. NC9745852 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0754020
This is an economical speed drive with seven speeds for reliable general transfer and feed applications. The drive is stackable and ideal for lab, field or process feed applications. The drive is IP22-rated and protected from water falling vertically.