Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, Similar to SC0461, Peroxide-Cured Silicone, 2.79 mm ID; 50 ft
Catalog No. NC1422737
Encompass
Reduce the risk of particulate entrapment with smooth inner surface. Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility. Global market leader in range of formulations for most extensive chemical compatibility.
Catalog No. NC1422737 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0762548
Reduce the risk of particulate entrapment with smooth inner surface. Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility. Global market leader in range of formulations for most extensive chemical compatibility.