Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Digital Modular Drive with Easy-Load Pump Head for Precision Tubing, 6 to 650 rpm; 115/230 VAC
Catalog No. NC1320843
Encompass
I/P modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation¿ideal for metering and critical transfer applications.
Catalog No. NC1320843 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7796220
Add to Cart
Add to Cart
Request a Quote
I/P modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation¿ideal for metering and critical transfer applications.