Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Digital Drive with Easy-Load II Pump Head for High-Performance Tubing, 600 rpm; 115/230 VAC
Catalog No. NC0623391
Encompass
These precision drives offer analog remote control options and simple programming¿ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy¿drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Catalog No. NC0623391 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7792175
These precision drives offer analog remote control options and simple programming¿ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy¿drive stores one calibration value per tubing size. Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.