Learn More
Masterflex™ L/S™ Easy-Load™ II Pump Heads
Catalog No. 13200026
Change view
Click to view available options
Flow Rate:
0.0012 to 2300 mL/min.
0.034 to 2900 mL/min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
13-200-026 0.034 to 2900 mL/min.
13-200-025 0.0012 to 2300 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-200-026 Supplier Masterflex™ Supplier No. MFLX7720062
Add to Cart
Edge
Add to Cart
Masterflex™ L/S™ Easy-Load™ II Pump Heads features automatic retention that holds tubing securely in place with no manual adjustment—just load the tubing and start the pump
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
Specifications
Description Fixed occlusion; No open-head sensor
Flow Rate 0.034 to 2900 mL/min.
For Use With High-Performance Precision Tubing
Includes Single-channel mounting hardware. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
Material PPS Housing, Stainless Steel Rotor
Tubing Type Tubing Sizes Accepted: L/S 15, 24, 35, 36