Learn More
Masterflex™ L/S™ Easy-Load™ Pump Heads for Precision Tubing
Description
Masterflex™ L/S™ Easy-Load™ Pump Heads for Precision Tubing saves time with fast tubing changes and reduced maintenance
Specifications
Specifications
| Flow Rate | 0.0012 to 2300 mL/min |
| Includes | Pump head |
| Material | Polyarylamide (PARA) housing, stainless steel rotor |
| Sensors | No open-head sensor |
| For Use With (Equipment) | Tubing sizes: L/S 13, 14, 16, 25, 17, 18 |