Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load II Pump Head with Adjustable Occlusion for Precision Tubing, PPS Housing, SS Rotor
Catalog No. NC0979384
Encompass
These pump heads accept the six L/S precision tubing sizes. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
Catalog No. NC0979384 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720160
These pump heads accept the six L/S precision tubing sizes. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load II pump heads are stacked rollers are offset by 45¿--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.