Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Analog, Variable-Speed, Modular Drive with Easy-Load 3 Pump Head, 6 to 600 rpm, 115 VAC
Catalog No. NC1398423
Encompass
Choose a convenient, complete system all with one catalog number. A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range and ¿2% speed regulation.
Catalog No. NC1398423 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791302
Choose a convenient, complete system all with one catalog number. A perfect upgrade from a fixed-speed drive, these variable-speed drives offer a wider rpm range and ¿2% speed regulation.