Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable Speed Analog Console Pump with Easy-Load 3 Pump Head for Precision Tubing, 6 to 600 rpm; 90 to 260 VAC
Catalog No. NC1496267
Encompass
These L/S precision variable-speed console drives offer precise ¿0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Catalog No. NC1496267 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791610
These L/S precision variable-speed console drives offer precise ¿0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.