Learn More
AVANTOR MASTERFLEX (VWR LLC) Spooled platinum-cured silicone tubing, L/S 16, 500 ft.
Catalog No. NC1589016
Encompass
Masterflex L/S Spooled Platinum-Cured Silicone Tubing, L/S 16, 500 ft, Slightly greater claritySmooth surface; low protein binding levelsPreferred by pharmaceutical & biotechnology industriesFewer potential leachablesUSP Class VI,
Catalog No. NC1589016 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640316/DEL
Masterflex L/S Spooled Platinum-Cured Silicone Tubing, L/S 16, 500 ft, Slightly greater claritySmooth surface; low protein binding levelsPreferred by pharmaceutical & biotechnology industriesFewer potential leachablesUSP Class VI,