Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Process Drive with Open-Head Sensor and Easy-Load II Pump Head for Precision Tubing, Stainless Steel Housing, 6 to 600 rpm; 90 to 260 VAC
Catalog No. NC1284485
Encompass
These accurate console drives have powerful motors, speed display, and ¿0.25% speed control for precise flow regulation¿ideal for metering applications. Remote speed control requires cable assembly 77300-32.
Catalog No. NC1284485 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791640
Add to Cart
Add to Cart
Request a Quote
These accurate console drives have powerful motors, speed display, and ¿0.25% speed control for precise flow regulation¿ideal for metering applications. Remote speed control requires cable assembly 77300-32.