Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable Speed Analog Console Pump with Easy-Load II Pump Head for High-Performance Tubing, 6 to 600 rpm; 90 to 260 VAC
Catalog No. NC0203722
Encompass
These L/S precision variable-speed console drives offer precise ¿0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.
Catalog No. NC0203722 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791620
These L/S precision variable-speed console drives offer precise ¿0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.