Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Fixed-Speed Drive, 12 rpm; 115 VAC
Catalog No. NC0563289
Encompass
The Masterflex L/S economy fixed-speed drives are compact and ideal where space is limited. These drives feature a lighted power switch on the front of the drive that lets you know when the pump is running. They accept five different pump head types: Standard, Easy-Load, Easy-Load II, Easy-Load 3, and Cartridge pump heads.
Catalog No. NC0563289 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0754012
The Masterflex L/S economy fixed-speed drives are compact and ideal where space is limited. These drives feature a lighted power switch on the front of the drive that lets you know when the pump is running. They accept five different pump head types: Standard, Easy-Load, Easy-Load II, Easy-Load 3, and Cartridge pump heads.