Learn More
Masterflex™ Accessories for Peristaltic Pump Systems
Catalog No. 11747359
Change view
Click to view available options
For Use With:
L/S digital drives
Catalog No. For Use With
11-747-359 L/S digital drives
1 options
Catalog No. 11-747-359 Supplier Masterflex™ Supplier No. MFLX0752392
Add to Cart
Add to Cart
Upgrade pumping and dispensing capabilities with Accessories for Masterflex™ Peristaltic Pump Systems.
  • Accessory items for the Masterflex™ Peristaltic Pump Systems
Note: Image is representative only; connection port may vary.  
Specifications
Description Masterflex Momentary Start/stop Foot Switch
For Use With (Application) Fluid handling
For Use With L/S digital drives
Includes DB25 connector, 1.8 m (6 ft.) cable
Quantity 1 Ea.
Speed Control Analog
Type Foot Switch