Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Sanitary Clamp with White Knob; 3/4""
Catalog No. NC1645925
Encompass
This clamp is compatible with ASME-BPE and BPSA standard nonmetallic and stainless steel flange ferrules. The lightweight design puts less stress on your system
Catalog No. NC1645925 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3120143
This clamp is compatible with ASME-BPE and BPSA standard nonmetallic and stainless steel flange ferrules. The lightweight design puts less stress on your system