Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Bulk-Packed Precision Pump Tubing, Platinum-Cured Silicone, L/S 16; 20 Bags of 25 ft/Bg
Catalog No. NC0320140
Encompass
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.
Catalog No. NC0320140 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640416/DEL
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.