Learn More
AVANTOR MASTERFLEX (VWR LLC) Polypropylene STRAIGHT,1/4 x 1/4 barb,10/Pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3061255/DEL
Encompass
Masterflex Polypropylene STRAIGHT,L/S 17/24 & I/P26, 1/4 x 1/4 barb, 10/PkSterilization Method- Autoclave or ethylene oxide (ETO)
Catalog No. NC1215567