Learn More
AVANTOR MASTERFLEX (VWR LLC) Multichannel Pump Head for Precision Tubing, 4-Channel
Catalog No. NC1830194
Encompass
Get low-pulsation synchronous flow in multiple channels without cartridges. Flow range 0.045 to 2300 mL/min per channel-flow rate depends on drive rpm and tubing size. Stainless steel (SS) rotor / roller for greater durability and chemical resistance.
Catalog No. NC1830194 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0753504
Get low-pulsation synchronous flow in multiple channels without cartridges. Flow range 0.045 to 2300 mL/min per channel-flow rate depends on drive rpm and tubing size. Stainless steel (SS) rotor / roller for greater durability and chemical resistance.