Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Sanitary Gasket, Buna-N, Flanged, 1-1/2"" Tri-Clamp; 1/EA
Catalog No. NC1795066
Encompass
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port¿once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time. Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).
Catalog No. NC1795066 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3056444
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port¿once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time. Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).