Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex B/T PerfertPosition Pump Tubing, Peroxide-Cured Silicone, B/T 90; 10 ft
Catalog No. NC0111646
Encompass
Ensure optimum performance from your Masterflex pump thanks to excellent biocompatibility of this tubing. No additives, plasticizers or DOP; odorless and nontoxic, fungus-resistant. No taste imparted to transported fluids. Extremely good at low temperatures. Weather, ozone, corona, and radiation resistant.
Catalog No. NC0111646 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9640090
Ensure optimum performance from your Masterflex pump thanks to excellent biocompatibility of this tubing. No additives, plasticizers or DOP; odorless and nontoxic, fungus-resistant. No taste imparted to transported fluids. Extremely good at low temperatures. Weather, ozone, corona, and radiation resistant.