Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P High-Performance Precision Pump Tubing, Platinum-Cured Silicone, I/P 89; 10 ft
Catalog No. NC0374923
Encompass
From a global market leader in range of formations, this tubing is distinguished by a smooth inner surface designed to reduce the risk of particle entrapment and contamination. Offering extensive chemical compatibility, genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.
Catalog No. NC0374923 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9651089/DEL
From a global market leader in range of formations, this tubing is distinguished by a smooth inner surface designed to reduce the risk of particle entrapment and contamination. Offering extensive chemical compatibility, genuine Masterflex tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment. Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility.